Anti-PSD4 Rabbit Polyclonal Antibody
Supplier: AFG Bioscience
New Product
Certificates
About this item
Recommended dilution: IHC:1:20-1:200
Type:
Antigen: PSD4
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77723-189
- 77723-190
- Antigen Symbol:
- PSD4
- PSD4
- Antigen Synonyms:
- Ppleckstrin and Sec7 domain containing 4|FLJ36237|Pleckstrin homology and SEC7 domain-containing protein 4|PSD4|PH and SEC7 domain-containing protein 4|PSD4_HUMAN|Exchange factor for ADP-ribosylation factor guanine nucleotide factor 6B|SEC7 homolog | telomeric of interleukin-1 cluster protein|TIC|EFA6B|FLJ37279|Telomeric of interleukin-1 cluster protein
- Ppleckstrin and Sec7 domain containing 4|FLJ36237|Pleckstrin homology and SEC7 domain-containing protein 4|PSD4|PH and SEC7 domain-containing protein 4|PSD4_HUMAN|Exchange factor for ADP-ribosylation factor guanine nucleotide factor 6B|SEC7 homolog | telomeric of interleukin-1 cluster protein|TIC|EFA6B|FLJ37279|Telomeric of interleukin-1 cluster protein
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q8NDX1
- Q8NDX1
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- Immunogen:
- Recombinant Human PH and SEC7 domain-containing protein 4 protein (1-300AA)
- Recombinant Human PH and SEC7 domain-containing protein 4 protein (1-300AA)
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on Blue Ice
- Shipped on Blue Ice
Specifications
Frequently Bought Together