Anti-ATG16L2 Rabbit Polyclonal Antibody (FITC)
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: ATG16L2
Clonality: Polyclonal
Clone:
Conjugation: FITC
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77723-140
- 77723-141
- Antigen Symbol:
- ATG16L2
- ATG16L2
- Antigen Synonyms:
- autophagy related 16-like 2 (S. cerevisiae)|Atg16l2|ATG16B|A16L2_HUMAN|APG16-like 2|Autophagy-related protein 16-2|WDR80|WD repeat-containing protein 80|ATG16 autophagy related 16-like 2
- autophagy related 16-like 2 (S. cerevisiae)|Atg16l2|ATG16B|A16L2_HUMAN|APG16-like 2|Autophagy-related protein 16-2|WDR80|WD repeat-containing protein 80|ATG16 autophagy related 16-like 2
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- FITC (Fluorescein Isothiocyanate)
- FITC (Fluorescein Isothiocyanate)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q8NAA4
- Q8NAA4
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Immunogen:
- Recombinant Human Autophagy-related protein 16-2 protein (262-348AA)
- Recombinant Human Autophagy-related protein 16-2 protein (262-348AA)
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on Blue Ice
- Shipped on Blue Ice
Specifications
Frequently Bought Together