Anti-PAF1 Rabbit Polyclonal Antibody (FITC)
New Product
Type:
Antigen: PAF1
Clonality: Polyclonal
Clone:
Conjugation: FITC
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
- Catalog No:
- 77723-081
- 77723-080
- Antigen Symbol:
- PAF1
- PAF1
- Antigen Synonyms:
- hPAF1|PAF 1|PD2 protein|RNA polymerase II-associated factor 1 homolog|PAF1_HUMAN|Pancreatic differentiation protein 2|FLJ11123|PD 2|PAF1|RNA polymerase II associated factor homolog |PD2|Paf1 RNA polymerase II associated factor homolog|F23149 1
- hPAF1|PAF 1|PD2 protein|RNA polymerase II-associated factor 1 homolog|PAF1_HUMAN|Pancreatic differentiation protein 2|FLJ11123|PD 2|PAF1|RNA polymerase II associated factor homolog |PD2|Paf1 RNA polymerase II associated factor homolog|F23149 1
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- FITC (Fluorescein Isothiocyanate)
- FITC (Fluorescein Isothiocyanate)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q8N7H5
- Q8N7H5
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Immunogen:
- Recombinant Human RNA polymerase II-associated factor 1 homolog protein (1-300AA)
- Recombinant Human RNA polymerase II-associated factor 1 homolog protein (1-300AA)
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on Blue Ice
- Shipped on Blue Ice
Frequently Bought Together