Anti-ARFGAP1 Rabbit Polyclonal Antibody
Supplier: AFG Bioscience
New Product
Certificates
About this item
Recommended dilution: WB:1:500-1:2000, IHC:1:20-1:200
Type:
Antigen: ARFGAP1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human; Mouse
Specifications
- Catalog No:
- 77723-075
- 77723-074
- Antigen Symbol:
- ARFGAP1
- ARFGAP1
- Antigen Synonyms:
- ARF1GAP|ARFG1_HUMAN|ADP-ribosylation factor 1 GTPase-activating protein|ADP-ribosylation factor GTPase-activating protein 1|ADP ribosylation factor GTPase activating protein 1|Arfgap1|GAP protein|ARF GAP 1|HRIHFB2281|ARF1 GAP|ARF1-directed GTPase-activating protein|ADP ribosylation factor 1 GTPase activating protein|ARF1 directed GTPase activating protein
- ARF1GAP|ARFG1_HUMAN|ADP-ribosylation factor 1 GTPase-activating protein|ADP-ribosylation factor GTPase-activating protein 1|ADP ribosylation factor GTPase activating protein 1|Arfgap1|GAP protein|ARF GAP 1|HRIHFB2281|ARF1 GAP|ARF1-directed GTPase-activating protein|ADP ribosylation factor 1 GTPase activating protein|ARF1 directed GTPase activating protein
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human, Mouse
- Human, Mouse
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q8N6T3
- Q8N6T3
- Isotype:
- IgG
- IgG
- Western Blot:
- Yes
- Yes
- ELISA:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- Immunogen:
- Recombinant Human ADP-ribosylation factor GTPase-activating protein 1 protein (1-220AA)
- Recombinant Human ADP-ribosylation factor GTPase-activating protein 1 protein (1-220AA)
- Form:
- Liquid
- Liquid
- Purification:
- Antigen affinity purified
- Antigen affinity purified
- Storage Buffer:
- PBS, 0.02% sodium azide, 50% glycerol, pH 7.3.
- PBS, 0.02% sodium azide, 50% glycerol, pH 7.3.
- Storage Temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on Blue Ice
- Shipped on Blue Ice
Specifications
Frequently Bought Together