Anti-NIN Rabbit Polyclonal Antibody (FITC)
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: NIN
Clonality: Polyclonal
Clone:
Conjugation: FITC
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77723-013
- 77723-012
- Antigen Symbol:
- NIN
- NIN
- Antigen Synonyms:
- NIN protein|ninein (GSK3B interacting protein)|SCKL7|h Glycogen synthase kinase 3 beta-interacting protein|NIN|Glycogen synthase kinase 3 beta interacting protein|KIAA1565|Ninein|NIN_HUMAN|Centrosome Marker|Ninein centrosomal protein|hNinein|GSK3B interacting protein
- NIN protein|ninein (GSK3B interacting protein)|SCKL7|h Glycogen synthase kinase 3 beta-interacting protein|NIN|Glycogen synthase kinase 3 beta interacting protein|KIAA1565|Ninein|NIN_HUMAN|Centrosome Marker|Ninein centrosomal protein|hNinein|GSK3B interacting protein
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- FITC (Fluorescein Isothiocyanate)
- FITC (Fluorescein Isothiocyanate)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q8N4C6
- Q8N4C6
- Isotype:
- IgG
- IgG
- Immunogen:
- Recombinant Human Ninein protein (80-185AA)
- Recombinant Human Ninein protein (80-185AA)
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on Blue Ice
- Shipped on Blue Ice
Specifications
Frequently Bought Together