Anti-ZFP82 Rabbit Polyclonal Antibody
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: ZFP82
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77722-963
- 77722-962
- Antigen Symbol:
- ZFP82
- ZFP82
- Antigen Synonyms:
- Zfp-82|Zinc finger protein 82 homolog|Zfp82|Zinc finger protein 545|ZFP82_HUMAN|Kiaa1948
- Zfp-82|Zinc finger protein 82 homolog|Zfp82|Zinc finger protein 545|ZFP82_HUMAN|Kiaa1948
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q8N141
- Q8N141
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Immunogen:
- Recombinant Human Zinc finger protein 82 homolog protein (1-200AA)
- Recombinant Human Zinc finger protein 82 homolog protein (1-200AA)
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on Blue Ice
- Shipped on Blue Ice
Specifications
Frequently Bought Together