Anti-NLGN4X Rabbit Polyclonal Antibody
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: NLGN4X
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77722-946
- 77722-947
- Antigen Symbol:
- NLGN4X
- NLGN4X
- Antigen Synonyms:
- KIAA1260|OTTHUMP00000022863|HNL4X|OTTHUMP00000022865|OTTHUMP00000022864|NLGNX_HUMAN|neuroligin 4, X-linked|Neuroligin-4|X-linked|HNLX|NLGN4X|NLGN4|NL4|Neuroligin X|ASPGX2|AUTSX2|NLGN
- KIAA1260|OTTHUMP00000022863|HNL4X|OTTHUMP00000022865|OTTHUMP00000022864|NLGNX_HUMAN|neuroligin 4, X-linked|Neuroligin-4|X-linked|HNLX|NLGN4X|NLGN4|NL4|Neuroligin X|ASPGX2|AUTSX2|NLGN
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q8N0W4
- Q8N0W4
- Isotype:
- IgG
- IgG
- Immunogen:
- Recombinant Human Neuroligin-4, X-linked protein (383-494AA)
- Recombinant Human Neuroligin-4, X-linked protein (383-494AA)
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on Blue Ice
- Shipped on Blue Ice
Specifications
Frequently Bought Together