Order Entry
United States
ContactUsLinkComponent
Anti-MED15 Rabbit Polyclonal Antibody (FITC)
Anti-MED15 Rabbit Polyclonal Antibody (FITC)
Supplier:  AFG Bioscience
New Product
Certificates
undefined
Anti-MED15 Rabbit Polyclonal Antibody (FITC)
Supplier:  AFG Bioscience
New Product

Specifications

  • Catalog No:
  • 77722-909
  • 77722-908
  • Antigen Symbol:
  • MED15
  • MED15
  • Antigen Synonyms:
  • CTG7A|PC2 glutamine/Q-rich-associated protein|TNRC7|TPA-inducible gene 1 protein|ARC105|Trinucleotide repeat-containing gene 7 protein|MED15|CTG repeat protein 7a|Mediator of RNA polymerase II transcription subunit 15|Activator-recruited cofactor 105 kDa component|Positive cofactor 2 glutamine/Q-rich-associated protein|TIG-1|PCQAP|TIG1|Mediator complex subunit 15
  • CTG7A|PC2 glutamine/Q-rich-associated protein|TNRC7|TPA-inducible gene 1 protein|ARC105|Trinucleotide repeat-containing gene 7 protein|MED15|CTG repeat protein 7a|Mediator of RNA polymerase II transcription subunit 15|Activator-recruited cofactor 105 kDa component|Positive cofactor 2 glutamine/Q-rich-associated protein|TIG-1|PCQAP|TIG1|Mediator complex subunit 15
  • Clonality:
  • Polyclonal
  • Polyclonal
  • Conjugation:
  • FITC (Fluorescein Isothiocyanate)
  • FITC (Fluorescein Isothiocyanate)
  • Reactivity:
  • Human
  • Human
  • Host:
  • Rabbit
  • Rabbit
  • Gene ID:
  • Q96RN5
  • Q96RN5
  • Isotype:
  • IgG
  • IgG
  • ELISA:
  • Yes
  • Yes
  • Immunogen:
  • Recombinant Human Mediator of RNA polymerase II transcription subunit 15 protein (433-530AA)
  • Recombinant Human Mediator of RNA polymerase II transcription subunit 15 protein (433-530AA)
  • Form:
  • Liquid
  • Liquid
  • Purification:
  • >95%, Protein G purified
  • >95%, Protein G purified
  • Storage Buffer:
  • 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
  • 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
  • Storage Temperature:
  • Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
  • Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
  • Shipping Temperature:
  • Shipped on Blue Ice
  • Shipped on Blue Ice

Specifications

Frequently Bought Together