Anti-C9orf72 Rabbit Polyclonal Antibody
Supplier: AFG Bioscience
New Product
Certificates
About this item
Recommended dilution: IHC:1:20-1:200, IF:1:50-1:200
Type:
Antigen: C9orf72
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77722-762
- 77722-761
- Antigen Symbol:
- C9orf72
- C9orf72
- Antigen Synonyms:
- RP11-27J8.2|chromosome 9 open reading frame 72|ALSFTD|C9orf72|Uncharacterized protein C9orf72|CI072_HUMAN|Protein C9orf72|MGC23980|FTDALS
- RP11-27J8.2|chromosome 9 open reading frame 72|ALSFTD|C9orf72|Uncharacterized protein C9orf72|CI072_HUMAN|Protein C9orf72|MGC23980|FTDALS
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q96LT7
- Q96LT7
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- ImmunoFluorescence:
- Yes
- Yes
- Immunogen:
- Recombinant Human Guanine nucleotide exchange C9orf72 protein (138-430AA)
- Recombinant Human Guanine nucleotide exchange C9orf72 protein (138-430AA)
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on Blue Ice
- Shipped on Blue Ice
Specifications
Frequently Bought Together