Anti-SIGLEC10 Rabbit Polyclonal Antibody (FITC)
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: SIGLEC10
Clonality: Polyclonal
Clone:
Conjugation: FITC
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77722-757
- 77722-758
- Antigen Symbol:
- SIGLEC10
- SIGLEC10
- Antigen Synonyms:
- sialic acid binding Ig like lectin 10 Ig like lectin 7|Siglecg|Sialic acid binding Immunoglobulin like lectin 10|Siglec G|Siglec like protein 2|SIG10_HUMAN|siglec like gene 2|PRO940|SIGLEC10|SLG2|sialic acid binding Ig like lectin G|Sialic acid binding Ig like lectin 10|Sialic acid-binding Ig-like lectin 10|Siglec-like protein 2|mSiglec G|SIGLEC 10|Siglec-10
- sialic acid binding Ig like lectin 10 Ig like lectin 7|Siglecg|Sialic acid binding Immunoglobulin like lectin 10|Siglec G|Siglec like protein 2|SIG10_HUMAN|siglec like gene 2|PRO940|SIGLEC10|SLG2|sialic acid binding Ig like lectin G|Sialic acid binding Ig like lectin 10|Sialic acid-binding Ig-like lectin 10|Siglec-like protein 2|mSiglec G|SIGLEC 10|Siglec-10
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- FITC (Fluorescein Isothiocyanate)
- FITC (Fluorescein Isothiocyanate)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q96LC7
- Q96LC7
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Immunogen:
- Recombinant Human Sialic acid-binding Ig-like lectin 10 protein (572-697AA)
- Recombinant Human Sialic acid-binding Ig-like lectin 10 protein (572-697AA)
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on Blue Ice
- Shipped on Blue Ice
Specifications
Frequently Bought Together