Anti-SPG11 Rabbit Polyclonal Antibody (HRP)
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: SPG11
Clonality: Polyclonal
Clone:
Conjugation: HRP
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77722-724
- 77722-725
- Antigen Symbol:
- SPG11
- SPG11
- Antigen Synonyms:
- DKFZp762B1512|SPG 11|FLJ21439|Spatacsin|Colorectal carcinoma associated protein|SPTCS_HUMAN|KIAA1840|Spastic paraplegia 11 (autosomal recessive)|Spastic paraplegia 11 protein|Colorectal carcinoma-associated protein|Spg11|Spastic paraplegia 11
- DKFZp762B1512|SPG 11|FLJ21439|Spatacsin|Colorectal carcinoma associated protein|SPTCS_HUMAN|KIAA1840|Spastic paraplegia 11 (autosomal recessive)|Spastic paraplegia 11 protein|Colorectal carcinoma-associated protein|Spg11|Spastic paraplegia 11
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- HRP (Horseradish Peroxidase)
- HRP (Horseradish Peroxidase)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q96JI7
- Q96JI7
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Immunogen:
- Recombinant Human Spatacsin protein (2019-2318AA)
- Recombinant Human Spatacsin protein (2019-2318AA)
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on Blue Ice
- Shipped on Blue Ice
Specifications
Frequently Bought Together