Anti-YIPF5 Rabbit Polyclonal Antibody (FITC)
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: YIPF5
Clonality: Polyclonal
Clone:
Conjugation: FITC
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77722-496
- 77722-495
- Antigen Symbol:
- YIPF5
- YIPF5
- Antigen Synonyms:
- SMAP 5|SMAP-5|SB140|YIPF5_HUMAN|Smooth muscle cell associated protein 5|FinGER5|SMAP5|yipf5|Five-pass transmembrane protein localizing in the Golgi apparatus and the endoplasmic reticulum 5|YPT-interacting protein 1 A|Five pass transmembrane protein localizing in the Golgi apparatus and the endoplasmic reticulum 5|Yip1 domain family member 5|YPT interacting protein 1 A|Smooth muscle cell-associated protein 5|YIP1A|Golgi membrane protein SB140|YIP1 family member 5|Protein YIPF5
- SMAP 5|SMAP-5|SB140|YIPF5_HUMAN|Smooth muscle cell associated protein 5|FinGER5|SMAP5|yipf5|Five-pass transmembrane protein localizing in the Golgi apparatus and the endoplasmic reticulum 5|YPT-interacting protein 1 A|Five pass transmembrane protein localizing in the Golgi apparatus and the endoplasmic reticulum 5|Yip1 domain family member 5|YPT interacting protein 1 A|Smooth muscle cell-associated protein 5|YIP1A|Golgi membrane protein SB140|YIP1 family member 5|Protein YIPF5
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- FITC (Fluorescein Isothiocyanate)
- FITC (Fluorescein Isothiocyanate)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q969M3
- Q969M3
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Immunogen:
- Recombinant Human Protein YIPF5 protein (1-124AA)
- Recombinant Human Protein YIPF5 protein (1-124AA)
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on Blue Ice
- Shipped on Blue Ice
Specifications
Frequently Bought Together