Anti-HOXB13 Rabbit Polyclonal Antibody (Biotin)
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: HOXB13
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77722-453
- 77722-454
- Antigen Symbol:
- HOXB13
- HOXB13
- Antigen Synonyms:
- PSGD protein|Homeobox protein HoxB13|Homeobox protein Hox-B13|PSGD|HXB13_HUMAN|HOX B13|Homeobox protein Hox B13|Homeobox B13|Homeo box B13|HOXB 13|HOXB13
- PSGD protein|Homeobox protein HoxB13|Homeobox protein Hox-B13|PSGD|HXB13_HUMAN|HOX B13|Homeobox protein Hox B13|Homeobox B13|Homeo box B13|HOXB 13|HOXB13
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- Biotin
- Biotin
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q92826
- Q92826
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Immunogen:
- Recombinant Human Homeobox protein Hox-B13 protein (1-161AA)
- Recombinant Human Homeobox protein Hox-B13 protein (1-161AA)
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on Blue Ice
- Shipped on Blue Ice
Specifications
Frequently Bought Together