Anti-TSEN15 Rabbit Polyclonal Antibody (FITC)
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: TSEN15
Clonality: Polyclonal
Clone:
Conjugation: FITC
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77722-290
- 77722-291
- Antigen Symbol:
- TSEN15
- TSEN15
- Antigen Synonyms:
- C1orf19|HsSEN15|SEN15|SEN15 homolog|tRNA-intron endonuclease Sen15|tRNA-splicing endonuclease subunit Sen15|TSEN15
- C1orf19|HsSEN15|SEN15|SEN15 homolog|tRNA-intron endonuclease Sen15|tRNA-splicing endonuclease subunit Sen15|TSEN15
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- FITC (Fluorescein Isothiocyanate)
- FITC (Fluorescein Isothiocyanate)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q8WW01
- Q8WW01
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Immunogen:
- Recombinant Human tRNA-splicing endonuclease subunit Sen15 protein (1-171AA)
- Recombinant Human tRNA-splicing endonuclease subunit Sen15 protein (1-171AA)
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on Blue Ice
- Shipped on Blue Ice
Specifications
Frequently Bought Together