Anti-MAPT (phospho–Thr231) Rabbit Polyclonal Antibody
Catalog # 77720-265
Supplier: AFG Bioscience
New Product
Specifications
- Antigen Symbol:MAPT (phospho-Thr231)
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western Blot:Yes
- Size:100 μl
- Form:liquid
- Gene ID:P10636
- Antigen Synonyms:MGC138549|PPP1R103|FTDP 17|Microtubule associated protein tau isoform 4|MAPT|AI413597|TAU_HUMAN|TAU|Paired helical filament-tau|Protein phosphatase 1, regulatory subunit 103|MGC134287|FLJ31424|PPND|Mtapt|MAPTL|G protein beta1/gamma2 subunit interacting factor 1|MTBT1|Tauopathy and respiratory failure|Tauopathy and respiratory failure, included|MTBT2|MGC156663|PHF tau|AW045860|Microtubule-associated protein tau|Neurofibrillary tangle protein|Microtubule associated protein tau|RNPTAU|pTau|Paired helical filament tau|PHF-tau|MSTD|DDPAC
- Storage Buffer:PBS, 150 mM NaCl, 0.02% sodium azide, 50% glycerol, pH 7.4.
- Storage Temperature:Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on Blue Ice
- Immunogen:Peptide sequence around phosphorylation site of threonine 231 (V-R-T(p)-P-P) derived from Human Tau.
- Purification:Antigen affinity purified
- Cat. No.:77720-265
- Packaging:Vial
Specifications
About this item
WB:1:500-1:1000, IHC:1:50-1:100
Type:
Antigen: MAPT (phospho–Thr231)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human; Mouse; Rat