Anti-HSF1 (phospho–Thr142) Rabbit Polyclonal Antibody
100 μl, Vial
Catalog # 77720-148
Supplier: AFG Bioscience
Supplier #: A62379-100UL
Jump to:
- Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
- Return Policy
Product Details & Documents
IHC:1:50-1:100
Type:
Antigen: HSF1 (phospho–Thr142)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human; Mouse
Specifications
- Size:100 μl
- Packaging:Vial
- Clonality:Polyclonal
- Purification:Antigen affinity purified
- Form:liquid
- Storage Buffer:PBS, 150 mM NaCl, 0.02% sodium azide, 50% glycerol, pH 7.4.
- Isotype:IgG
- Host:Rabbit
- Cat. No.:77720-148
- Reactivity:Human|Mouse
- Shipping Temperature:Shipped on Blue Ice
- Antigen Symbol:HSF1 (phospho-Thr142)
- Gene ID:Q00613
- Immunogen:Peptide sequence around phosphorylation site of threonine 142(L-L-T(p)-D-V) derived from Human HSF1 .
- ImmunoChemistry:Yes
- Conjugation:Unconjugated
- Antigen Synonyms:HSTF 1|Heat shock transcription factor 1|Heat shock factor protein 1|HSTF1|hsf1|HSF 1|HSF1_HUMAN|Heat shock factor 1
- ELISA:Yes
- Storage Temperature:Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



