Anti-USP50 Rabbit Polyclonal Antibody
Catalog # 77716-227
Supplier: AFG Bioscience
New Product
Specifications
- Antigen Symbol:USP50
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Mouse
- Western Blot:Yes
- Size:100 μl
- Form:liquid
- Gene ID:Q70EL3
- Antigen Synonyms:Inactive ubiquitin carboxyl-terminal hydrolase 50|Inactive ubiquitin-specific peptidase 50|USP50
- Storage Buffer:PBS, 150 mM NaCl, 0.02% sodium azide, 50% glycerol, pH 7.4.
- Storage Temperature:Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on Blue Ice
- Immunogen:Synthesized peptide derived from C-terminal of Human USP50.
- Purification:Antigen affinity purified
- Cat. No.:77716-227
- Packaging:Vial
Specifications
About this item
WB:1:500-1:3000, IHC:1:50-1:100
Type:
Antigen: USP50
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human; Mouse