Anti-MRPS12 Rabbit Polyclonal Antibody
Catalog # 77716-131
Supplier: AFG Bioscience
New Product
Specifications
- Antigen Symbol:MRPS12
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human
- Western Blot:Yes
- Size:100 μl
- Form:liquid
- Gene ID:O15235
- Antigen Synonyms:MT-RPS12|MRPS12|28S ribosomal protein S12|MRP-S12|mitochondrial|RPSM12|RPMS12|Mitochondrial ribosomal protein S12|S12mt|RPS12|28S ribosomal protein S12 mitochondrial|RT12_HUMAN
- Storage Buffer:PBS, 150 mM NaCl, 0.02% sodium azide, 50% glycerol, pH 7.4.
- Storage Temperature:Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on Blue Ice
- Immunogen:Synthesized peptide derived from internal of Human MRPS12.
- Purification:Antigen affinity purified
- Cat. No.:77716-131
- Packaging:Vial
Specifications
About this item
WB:1:500-1:3000
Type:
Antigen: MRPS12
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human