Anti-MACC1 Rabbit Polyclonal Antibody
77689-916
New Product
- Antigen Symbol:MACC1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human
- Western Blot:Yes
- Size:100 μg
- Form:Liquid
- Gene ID:Q6ZN28
- Antigen Synonyms:7A5|MACC1|Metastasis associated in colon cancer 1|SH3BP4L|SH3 domain-containing protein 7a5|MACC1_HUMAN|Metastasis-associated in colon cancer protein 1|OTTHUMP00000201891|Putative binding protein 7a5
- Storage Buffer:0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on Blue Ice
- Immunogen:Recombinant Human Metastasis-associated in colon cancer protein 1 protein (371-514AA)
- Purification:>95%, Protein G purified
- Cat. No.:77689-916
- Packaging:Vial
Recommended dilution: WB:1:500-1:5000, IHC:1:20-1:200, IF:1:50-1:200
Type:
Antigen: MACC1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human