Anti-PRKAG1 Rabbit Polyclonal Antibody
Catalog # 77705-596
Supplier: AFG Bioscience
New Product
Specifications
- Antigen Symbol:PRKAG1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western Blot:Yes
- Size:100 μg
- Form:Liquid
- Gene ID:P54619
- Antigen Synonyms:5''-AMP-activated protein kinase subunit gamma-1|MGC8666|noncatalytic gamma-1|Protein kinase AMP activated gamma 1 non catalytic subunit|AMPK subunit gamma-1|protein kinase|AMPKg|5' AMP activated protein kinase gamma 1 subunit|5' AMP activated protein kinase subunit gamma 1|AMPK gamma 1 chain|AAKG1_HUMAN|AMP activated protein kinase noncatalytic gamma 1 subunit|AMP-activated|PRKAG1|AMPK gamma1|PRKAG 1
- Storage Buffer:PBS, 50% glycerol, 0.5% BSA, 0.02% sodium azide, pH 7.4.
- Storage Temperature:Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on Blue Ice
- Immunogen:Synthesized peptide derived from the N-terminal region of Human AMPKγ1.
- Purification:Antigen affinity purified
- Cat. No.:77705-596
- Packaging:Vial
Specifications
About this item
WB:1:500-1:2000, ELISA:1:5000
Type:
Antigen: PRKAG1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human; Mouse; Rat