Anti-NUGGC Rabbit Polyclonal Antibody
Catalog # 77689-761
Supplier: AFG Bioscience
New Product
Specifications
- Antigen Symbol:NUGGC
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human
- Western Blot:Yes
- Size:100 μl
- Form:Liquid
- Gene ID:Q68CJ6
- Antigen Synonyms:NUGGC|Nuclear GTPase SLIP-GC|EC 3.6.1.-|C8orf80|Speckled-like pattern in the germinal center|HMFN0672
- Storage Buffer:0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on Blue Ice
- Immunogen:Recombinant Human Nuclear GTPase SLIP-GC protein (1-228AA)
- Purification:>95%, Protein G purified
- Cat. No.:77689-761
- Packaging:Vial
Specifications
About this item
Recommended dilution: WB:1:1000-1:5000
Type:
Antigen: NUGGC
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human