Anti-IGSF11 Rabbit Polyclonal Antibody
Catalog # 77689-725
Supplier: AFG Bioscience
New Product
Specifications
- Antigen Symbol:IGSF11
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Mouse
- Western Blot:Yes
- Size:100 μg
- Form:Liquid
- Gene ID:Q5DX21
- Antigen Synonyms:IGSF11|Brain and testis-specific immunoglobulin superfamily protein|IgSF11|Immunoglobulin superfamily member 11|BTIGSF|CXADRL1|Bt-IGSF|V-set and immunoglobulin domain-containing protein 3|VSIG3
- Storage Buffer:0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on Blue Ice
- Immunogen:Recombinant Human Immunoglobulin superfamily member 11 protein (23-241AA)
- Purification:Antigen affinity purified
- Cat. No.:77689-725
- Packaging:Vial
Specifications
About this item
Recommended dilution: WB:1:500-1:2000
Type:
Antigen: IGSF11
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human; Mouse