Anti-UNC50 Rabbit Polyclonal Antibody
Catalog # 77689-720
Supplier: AFG Bioscience
New Product
Specifications
- Antigen Symbol:UNC50
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human
- Western Blot:Yes
- Size:100 μg
- Form:Liquid
- Gene ID:Q53HI1
- Antigen Synonyms:Protein unc-50 homolog|UNCL|PDLs22|Periodontal ligament-specific protein 22|Uncoordinated-like protein|hGMH1|UNC50|HSD-23|HSD23|Protein GMH1 homolog
- Storage Buffer:0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on Blue Ice
- Immunogen:Recombinant Human Protein unc-50 homolog protein (15-82AA)
- Purification:Antigen affinity purified
- Cat. No.:77689-720
- Packaging:Vial
Specifications
About this item
Recommended dilution: WB:1:1000-1:5000, IF:1:50-1:200
Type:
Antigen: UNC50
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human