Anti-MRPL51 Rabbit Polyclonal Antibody
Catalog # 77689-717
Supplier: AFG Bioscience
New Product
Specifications
- Antigen Symbol:MRPL51
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human
- Western Blot:Yes
- Size:100 μg
- Form:Liquid
- Gene ID:Q4U2R6
- Antigen Synonyms:mitochondrial ribosomal protein L51|bMRP-64|HSPC241|MRP-L51|mitochondrial ribosomal protein 64|bMRP64|L51mt|mrpl51|mitochondrial|RM51_HUMAN|39S ribosomal protein L51 mitochondrial|MRP64|CDA09|mitochondrial ribosomal protein bMRP64|39S ribosomal protein L51
- Storage Buffer:0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on Blue Ice
- Immunogen:Recombinant Human 39S ribosomal protein L51, mitochondrial protein (32-128AA)
- Purification:>95%, Protein G purified
- Cat. No.:77689-717
- Packaging:Vial
Specifications
About this item
Recommended dilution: WB:1:500-1:2000, IHC:1:20-1:200
Type:
Antigen: MRPL51
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human