Anti-GFOD2 Rabbit Polyclonal Antibody
Catalog # 77689-694
Supplier: AFG Bioscience
New Product
Specifications
- Antigen Symbol:GFOD2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human
- Western Blot:Yes
- Size:100 μg
- Form:Liquid
- Gene ID:Q3B7J2
- Antigen Synonyms:Glucose fructose oxidoreductase domain containing 2|Glucose-fructose oxidoreductase domain-containing protein 2|MGC11335|Glucose fructose oxidoreductase domain containing protein 2|GFOD2_HUMAN|FLJ23802|GFOD2
- Storage Buffer:0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on Blue Ice
- Immunogen:Recombinant Human Glucose-fructose oxidoreductase domain-containing protein 2 protein (1-280AA)
- Purification:Antigen affinity purified
- Cat. No.:77689-694
- Packaging:Vial
Specifications
About this item
Recommended dilution: WB:1:500-1:5000, IHC:1:20-1:200
Type:
Antigen: GFOD2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human