Anti-SLC13A2 Rabbit Polyclonal Antibody
Catalog # 77689-576
Supplier: AFG Bioscience
New Product
Specifications
- Antigen Symbol:SLC13A2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human
- Western Blot:Yes
- Size:100 μl
- Form:Liquid
- Gene ID:Q13183
- Antigen Synonyms:NaDC-1|NADC1|SDCT1|SLC13A2|Renal sodium/dicarboxylate cotransporter|Na(+)/dicarboxylate cotransporter 1|Solute carrier family 13 member 2
- Storage Buffer:PBS, 0.02% sodium azide, 50% glycerol, pH 7.3.
- Storage Temperature:Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on Blue Ice
- Immunogen:Recombinant Human Solute carrier family 13 member 2 protein (345-455AA)
- Purification:Antigen affinity purified
- Cat. No.:77689-576
- Packaging:Vial
Specifications
About this item
Recommended dilution: WB:1:1000-1:5000, IHC:1:20-1:200
Type:
Antigen: SLC13A2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human