Anti-DUSP7 Rabbit Polyclonal Antibody
Catalog # 77689-522
Supplier: AFG Bioscience
New Product
Specifications
- Antigen Symbol:DUSP7
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human
- Western Blot:Yes
- Size:100 μl
- Form:Liquid
- Gene ID:Q16829
- Antigen Synonyms:Dual specificity protein phosphatase PYST2|Dusp7|JSP1|MKPX|Jnk Stimulatory Phosphatase 1|Dual specificity phosphatase 7|DUS7_HUMAN|MAP kinase phosphatase X|Dual specificity protein phosphatase 7|PYST2
- Storage Buffer:PBS, 0.02% sodium azide, 50% glycerol, pH 7.3.
- Storage Temperature:Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on Blue Ice
- Immunogen:Recombinant Human Dual specificity protein phosphatase 7 protein (160-419AA)
- Purification:Antigen affinity purified
- Cat. No.:77689-522
- Packaging:Vial
Specifications
About this item
Recommended dilution: WB:1:500-1:2000, IHC:1:20-1:200
Type:
Antigen: DUSP7
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human