Anti-ANGPT1 Rabbit Polyclonal Antibody
Supplier: AFG Bioscience
New Product
Certificates
About this item
ELISA:1:2000-1:5000, IHC:1:50-1:200
Type:
Antigen: ANGPT1
Clonality: Polyclonal
Clone:
Conjucation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Specifications
- Catalog No:
- 77714-718
- 77714-719
- Antigen Symbol:
- ANGPT1
- ANGPT1
- Antigen Synonyms:
- OTTHUMP00000227599|OTTHUMP00000227592|KIAA0003|ANG1|Angiopoietin-1|AGP1|Angpt1|OTTHUMP00000227600|ANG-1|ANGPT 1|ANG 1|AGP 1|ANGP1_HUMAN|Angiopoietin1|ANG|AGPT
- OTTHUMP00000227599|OTTHUMP00000227592|KIAA0003|ANG1|Angiopoietin-1|AGP1|Angpt1|OTTHUMP00000227600|ANG-1|ANGPT 1|ANG 1|AGP 1|ANGP1_HUMAN|Angiopoietin1|ANG|AGPT
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human, Rat, Mouse
- Human, Rat, Mouse
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q15389
- Q15389
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- Immunogen:
- Fusion protein of Human ANGPT1
- Fusion protein of Human ANGPT1
- Form:
- Liquid
- Liquid
- Purification:
- Antigen affinity purified
- Antigen affinity purified
- Storage Buffer:
- PBS, 0.05% sodium azide, 40% glycerol, pH 7.4.
- PBS, 0.05% sodium azide, 40% glycerol, pH 7.4.
- Storage Temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together