Anti-AANAT Rabbit Polyclonal Antibody
Supplier: AFG Bioscience
New Product
Certificates
About this item
ELISA:1:1000-1:5000, IHC:1:50-1:200
Type:
Antigen: AANAT
Clonality: Polyclonal
Clone:
Conjucation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Specifications
- Catalog No:
- 77714-549
- 77714-550
- Antigen Synonyms:
- Aanat|Serotonin N-acetyltransferase|Aralkylamine N-acetyltransferase|SNAT|Arylalkylamine N acetyltransferase|SNAT_HUMAN|AA-NAT|Serotonin acetylase
- Aanat|Serotonin N-acetyltransferase|Aralkylamine N-acetyltransferase|SNAT|Arylalkylamine N acetyltransferase|SNAT_HUMAN|AA-NAT|Serotonin acetylase
- Gene ID:
- Q16613
- Q16613
- Immunogen:
- Fusion protein of Human AANAT
- Fusion protein of Human AANAT
- Form:
- Liquid
- Liquid
- Purification:
- Antigen affinity purified
- Antigen affinity purified
- Storage Buffer:
- PBS, 0.05% sodium azide, 40% glycerol, pH 7.4.
- PBS, 0.05% sodium azide, 40% glycerol, pH 7.4.
- Storage Temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together