Anti-UBIAD1 Rabbit Polyclonal Antibody (HRP)
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: UBIAD1
Clonality: Polyclonal
Clone:
Conjucation: HRP
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77714-434
- 77714-433
- Antigen Symbol:
- UBIAD1
- UBIAD1
- Antigen Synonyms:
- UBIAD 1|UbiA prenyltransferase domain containing 1|TERE 1|Transitional epithelial response protein 1|Transitional epithelia response protein|UbiA prenyltransferase domain containing protein 1|RP4-796F18.1|UbiA prenyltransferase domain-containing protein 1|SCCD|Ubiad1|UBIA1_HUMAN|tere1
- UBIAD 1|UbiA prenyltransferase domain containing 1|TERE 1|Transitional epithelial response protein 1|Transitional epithelia response protein|UbiA prenyltransferase domain containing protein 1|RP4-796F18.1|UbiA prenyltransferase domain-containing protein 1|SCCD|Ubiad1|UBIA1_HUMAN|tere1
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- HRP (Horseradish Peroxidase)
- HRP (Horseradish Peroxidase)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q9Y5Z9
- Q9Y5Z9
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Immunogen:
- Recombinant Human UbiA prenyltransferase domain-containing protein 1 protein (1-82AA)
- Recombinant Human UbiA prenyltransferase domain-containing protein 1 protein (1-82AA)
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together