Anti-PDE1A Rabbit Polyclonal Antibody
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: PDE1A
Clonality: Polyclonal
Clone:
Conjucation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77714-237
- 77714-238
- Antigen Symbol:
- PDE1A
- PDE1A
- Antigen Synonyms:
- HCAM 1|Phosphodiesterase 1A calmodulin dependent|Calcium/calmodulin-dependent 3''|3' 5' cyclic nucleotide phosphodiesterase|61 kDa Cam-PDE|61 kDa Cam PDE|CAM-PDE-1A|MGC26303|Calcium/Calmodulin stimulated cyclic nucleotide phosphodiesterase|PDE1A_HUMAN|Calmodulin-dependent Phosphodiesterase|Phosphodiesterase 1A|Calcium/calmodulin dependent 3' 5' cyclic nucleotide phosphodiesterase 1A|HSPDE1A|5''-cyclic nucleotide phosphodiesterase 1A|PDE1A|HCAM1|hCam-1|PDE 1A|Cam-PDE 1A|Cam PDE 1A
- HCAM 1|Phosphodiesterase 1A calmodulin dependent|Calcium/calmodulin-dependent 3''|3' 5' cyclic nucleotide phosphodiesterase|61 kDa Cam-PDE|61 kDa Cam PDE|CAM-PDE-1A|MGC26303|Calcium/Calmodulin stimulated cyclic nucleotide phosphodiesterase|PDE1A_HUMAN|Calmodulin-dependent Phosphodiesterase|Phosphodiesterase 1A|Calcium/calmodulin dependent 3' 5' cyclic nucleotide phosphodiesterase 1A|HSPDE1A|5''-cyclic nucleotide phosphodiesterase 1A|PDE1A|HCAM1|hCam-1|PDE 1A|Cam-PDE 1A|Cam PDE 1A
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- P54750
- P54750
- Isotype:
- IgG
- IgG
- Immunogen:
- Recombinant Human Calcium/calmodulin-dependent 3\',5\'-cyclic nucleotide phosphodiesterase 1A protein (1-219AA)
- Recombinant Human Calcium/calmodulin-dependent 3\',5\'-cyclic nucleotide phosphodiesterase 1A protein (1-219AA)
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together