Anti-KIAA0319 Rabbit Polyclonal Antibody (FITC)
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: KIAA0319
Clonality: Polyclonal
Clone:
Conjucation: FITC
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77714-197
- 77714-196
- Antigen Symbol:
- KIAA0319
- KIAA0319
- Antigen Synonyms:
- DYX2|MGC176717|DYX 2|DLX2|DYLX 2|DYLX2|Dyslexia-associated protein KIAA0319|K0319_HUMAN|Kiaa0319|Dyslexia susceptibility 2|DLX 2
- DYX2|MGC176717|DYX 2|DLX2|DYLX 2|DYLX2|Dyslexia-associated protein KIAA0319|K0319_HUMAN|Kiaa0319|Dyslexia susceptibility 2|DLX 2
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- FITC (Fluorescein Isothiocyanate)
- FITC (Fluorescein Isothiocyanate)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q5VV43
- Q5VV43
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Immunogen:
- Recombinant Human Dyslexia-associated protein KIAA0319 protein (153-268AA)
- Recombinant Human Dyslexia-associated protein KIAA0319 protein (153-268AA)
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together