Anti-CLC Rabbit Polyclonal Antibody (HRP)
New Product
Type:
Antigen: CLC
Clonality: Polyclonal
Clone:
Conjucation: HRP
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
- Catalog No:
- 77714-183
- 77714-184
- Antigen Synonyms:
- LGALS10A|LPPL HUMAN|Galectin-10|Lysolecithin acylhydrolase|Gal-10|Eosinophil lysophospholipase|LGALS10|BSF3|EC 3.1.1.5|CLC|Charcot Leyden crystal protein|MGC149659|Charcot-Leyden crystal protein|LPPL_HUMAN
- LGALS10A|LPPL HUMAN|Galectin-10|Lysolecithin acylhydrolase|Gal-10|Eosinophil lysophospholipase|LGALS10|BSF3|EC 3.1.1.5|CLC|Charcot Leyden crystal protein|MGC149659|Charcot-Leyden crystal protein|LPPL_HUMAN
- Gene ID:
- Q05315
- Q05315
- Immunogen:
- Recombinant Human Galectin-10 protein (2-142AA)
- Recombinant Human Galectin-10 protein (2-142AA)
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Frequently Bought Together