Anti-CXCR6 Rabbit Polyclonal Antibody
Supplier: AFG Bioscience
New Product
Certificates
About this item
ELISA:1:2000-1:5000, WB:1:500-1:2000, IHC:1:50-1:200
Type:
Antigen: CXCR6
Clonality: Polyclonal
Clone:
Conjucation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Specifications
- Catalog No:
- 77714-029
- 77714-028
- Antigen Synonyms:
- Chemokine receptor 6|Chemokine CXC motif receptor 6|Chemokine C X C motif receptor 6|TYMSTR|CXC chemokine receptor type 6|bonzo|C-X-C chemokine receptor type 6|G-protein coupled receptor STRL33|CXC R6|STRL 33|CD186|CXCR6_HUMAN|STRL33|G protein coupled receptor bonzo|CD 186|CXCR6|CXCR-6|G protein coupled receptor|G-protein coupled receptor bonzo|G protein coupled receptor TYMSTR|CDw186|C X C chemokine receptor type 6|G protein coupled receptor STRL 33|CXCR 6|G protein coupled receptor STRL33|CXC-R6|Chemokine (C X C motif) receptor 6|CD186 antigen
- Chemokine receptor 6|Chemokine CXC motif receptor 6|Chemokine C X C motif receptor 6|TYMSTR|CXC chemokine receptor type 6|bonzo|C-X-C chemokine receptor type 6|G-protein coupled receptor STRL33|CXC R6|STRL 33|CD186|CXCR6_HUMAN|STRL33|G protein coupled receptor bonzo|CD 186|CXCR6|CXCR-6|G protein coupled receptor|G-protein coupled receptor bonzo|G protein coupled receptor TYMSTR|CDw186|C X C chemokine receptor type 6|G protein coupled receptor STRL 33|CXCR 6|G protein coupled receptor STRL33|CXC-R6|Chemokine (C X C motif) receptor 6|CD186 antigen
- Gene ID:
- O00574
- O00574
- Immunogen:
- Fusion protein of Human CXCR6
- Fusion protein of Human CXCR6
- Form:
- Liquid
- Liquid
- Purification:
- Antigen affinity purified
- Antigen affinity purified
- Storage Buffer:
- PBS, 0.05% sodium azide, 40% glycerol, pH 7.4.
- PBS, 0.05% sodium azide, 40% glycerol, pH 7.4.
- Storage Temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together