Anti-ELOVL6 Rabbit Polyclonal Antibody
Supplier: AFG Bioscience
New Product
Certificates
About this item
ELISA:1:2000-1:5000, WB:1:500-1:2000, IHC:1:25-1:100
Type:
Antigen: ELOVL6
Clonality: Polyclonal
Clone:
Conjucation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Specifications
- Catalog No:
- 77714-018
- 77714-017
- Antigen Synonyms:
- FLJ23378|Fatty acid elongase 2|Fatty acyl-CoA elongase|FACE|MGC5487|ELOVL family member 6|ELOVL fatty acid elongase 6|Long-chain fatty-acyl elongase|ELOV6_HUMAN|Long chain fatty acyl elongase|3-keto acyl-CoA synthase ELOVL6|Myelin associated SUR4 protein|Elongation of very long chain fatty acids protein 6|hELO2|SUR4/Elo3 like|Fatty acyl CoA elongase|elongation of long chain fatty acids (FEN1/Elo2|ELOVL6|FAE|yeast)|LCE
- FLJ23378|Fatty acid elongase 2|Fatty acyl-CoA elongase|FACE|MGC5487|ELOVL family member 6|ELOVL fatty acid elongase 6|Long-chain fatty-acyl elongase|ELOV6_HUMAN|Long chain fatty acyl elongase|3-keto acyl-CoA synthase ELOVL6|Myelin associated SUR4 protein|Elongation of very long chain fatty acids protein 6|hELO2|SUR4/Elo3 like|Fatty acyl CoA elongase|elongation of long chain fatty acids (FEN1/Elo2|ELOVL6|FAE|yeast)|LCE
- Gene ID:
- Q9H5J4
- Q9H5J4
- Immunogen:
- Synthetic peptide of Human ELOVL6
- Synthetic peptide of Human ELOVL6
- Form:
- Liquid
- Liquid
- Purification:
- Antigen affinity purified
- Antigen affinity purified
- Storage Buffer:
- PBS, 0.05% sodium azide, 40% glycerol, pH 7.4.
- PBS, 0.05% sodium azide, 40% glycerol, pH 7.4.
- Storage Temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together