Anti-RCC1 Rabbit Polyclonal Antibody
Supplier: AFG Bioscience
New Product
Certificates
About this item
ELISA:1:1000-1:2000, WB:1:200-1:1000
Type:
Antigen: RCC1
Clonality: Polyclonal
Clone:
Conjucation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Specifications
- Catalog No:
- 77713-942
- 77713-943
- Antigen Symbol:
- RCC1
- RCC1
- Antigen Synonyms:
- Chromosome condensation 1|HERC2|RCC1_HUMAN|RanGEF|CHC 1|SNHG3 RCC1 readthrough transcript|Cell cycle regulatory protein|RCC 1|Regulator of chromosome condensation|SNHG3 RCC1|Chromosome condensation protein 1|RCC1|Regulator of chromosome condensation 1|CHC1|SHEP1|Ran GEF|Guanine nucleotide releasing protein|RCC1 I
- Chromosome condensation 1|HERC2|RCC1_HUMAN|RanGEF|CHC 1|SNHG3 RCC1 readthrough transcript|Cell cycle regulatory protein|RCC 1|Regulator of chromosome condensation|SNHG3 RCC1|Chromosome condensation protein 1|RCC1|Regulator of chromosome condensation 1|CHC1|SHEP1|Ran GEF|Guanine nucleotide releasing protein|RCC1 I
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human, Mouse
- Human, Mouse
- Host:
- Rabbit
- Rabbit
- Gene ID:
- P18754
- P18754
- Isotype:
- IgG
- IgG
- Western Blot:
- Yes
- Yes
- ELISA:
- Yes
- Yes
- Immunogen:
- Synthetic peptide of Human RCC1
- Synthetic peptide of Human RCC1
- Form:
- Liquid
- Liquid
- Purification:
- Antigen affinity purified
- Antigen affinity purified
- Storage Buffer:
- PBS, 0.05% sodium azide, 40% glycerol, pH 7.4.
- PBS, 0.05% sodium azide, 40% glycerol, pH 7.4.
- Storage Temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together