Anti-SYT6 Rabbit Polyclonal Antibody
Supplier: AFG Bioscience
New Product
Certificates
About this item
ELISA:1:2000-1:5000, IHC:1:50-1:200
Type:
Antigen: SYT6
Clonality: Polyclonal
Clone:
Conjucation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Specifications
- Catalog No:
- 77713-766
- 77713-765
- Antigen Symbol:
- SYT6
- SYT6
- Antigen Synonyms:
- Synaptotagmin-6|Synaptotagmin VI|Synaptotagmin6|SYT6_HUMAN|Syt VI|SynaptotagminVI|Synaptotagmin 6|SytVI|SYT6|Syt 6
- Synaptotagmin-6|Synaptotagmin VI|Synaptotagmin6|SYT6_HUMAN|Syt VI|SynaptotagminVI|Synaptotagmin 6|SytVI|SYT6|Syt 6
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human, Rat, Mouse
- Human, Rat, Mouse
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q5T7P8
- Q5T7P8
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- Immunogen:
- Fusion protein of Human SYT6
- Fusion protein of Human SYT6
- Form:
- Liquid
- Liquid
- Purification:
- Antigen affinity purified
- Antigen affinity purified
- Storage Buffer:
- PBS, 0.05% sodium azide, 40% glycerol, pH 7.4
- PBS, 0.05% sodium azide, 40% glycerol, pH 7.4
- Storage Temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together