Order Entry
United States
ContactUsLinkComponent
Anti-XRCC6 (acetyl-K542) Rabbit Polyclonal Antibody
Anti-XRCC6 (acetyl-K542) Rabbit Polyclonal Antibody
Supplier:  AFG Bioscience
New Product
Certificates
undefined
Anti-XRCC6 (acetyl-K542) Rabbit Polyclonal Antibody
Supplier:  AFG Bioscience
New Product

Specifications

  • Catalog No:
  • 77713-684
  • 77705-354
  • Antigen Symbol:
  • XRCC6 (acetyl-K542)
  • XRCC6 (acetyl-K542)
  • Antigen Synonyms:
  • CTC box binding factor 75 kDa subunit|Kup70|ATP dependent DNA helicase II 70 kDa subunit|DNA repair protein XRCC6|XRCC 6|Ku 70|Ku autoantigen, 70kDa|Thyroid-lupus autoantigen|70 kDa subunit of Ku antigen|Ku autoantigen p70 subunit|XRCC6_HUMAN|TLAA|X-ray repair complementing defective repair in Chinese hamster cells 6|X ray repair complementing defective repair in Chinese hamster cells 6|Thyroid autoantigen 70kD (Ku antigen)|Ku70 DNA binding component of DNA-dependent proteinkinase complex (thyroid autoantigen 70 kDa|Thyroid lupus autoantigen|Xrcc6|CTC75|CTCBF|X-ray repair cross-complementing protein 6|ATP dependent DNA helicase 2 subunit 1|Ku70|CTC box-binding factor 75 kDa subunit|Thyroid lupus autoantigen p70|ATP-dependent DNA helicase 2 subunit 1|ML8|5''-dRP lyase Ku70|Lupus Ku autoantigen protein p70|Thyroid autoantigen|G22P1|5''-deoxyribose-5-phosphate lyase Ku70|ATP-dependent DNA helicase II 70 kDa subunit|Ku p70
  • CTC box binding factor 75 kDa subunit|Kup70|ATP dependent DNA helicase II 70 kDa subunit|DNA repair protein XRCC6|XRCC 6|Ku 70|Ku autoantigen, 70kDa|Thyroid-lupus autoantigen|70 kDa subunit of Ku antigen|Ku autoantigen p70 subunit|XRCC6_HUMAN|TLAA|X-ray repair complementing defective repair in Chinese hamster cells 6|X ray repair complementing defective repair in Chinese hamster cells 6|Thyroid autoantigen 70kD (Ku antigen)|Ku70 DNA binding component of DNA-dependent proteinkinase complex (thyroid autoantigen 70 kDa|Thyroid lupus autoantigen|Xrcc6|CTC75|CTCBF|X-ray repair cross-complementing protein 6|ATP dependent DNA helicase 2 subunit 1|Ku70|CTC box-binding factor 75 kDa subunit|Thyroid lupus autoantigen p70|ATP-dependent DNA helicase 2 subunit 1|ML8|5''-dRP lyase Ku70|Lupus Ku autoantigen protein p70|Thyroid autoantigen|G22P1|5''-deoxyribose-5-phosphate lyase Ku70|ATP-dependent DNA helicase II 70 kDa subunit|Ku p70
  • Clonality:
  • Polyclonal
  • Polyclonal
  • Conjugation:
  • Unconjugated
  • Unconjugated
  • Reactivity:
  • Human
  • Human
  • Host:
  • Rabbit
  • Rabbit
  • Gene ID:
  • P12956
  • P12956
  • Isotype:
  • IgG
  • IgG
  • Western Blot:
  • Yes
  • Yes
  • ELISA:
  • Yes
  • Yes
  • Immunogen:
  • Synthesized peptide derived from the Human Ku-70 around the acetylation site of K542.
  • Synthesized peptide derived from the Human Ku-70 around the acetylation site of K542.
  • Form:
  • Liquid
  • Liquid
  • Purification:
  • Antigen affinity purified
  • Antigen affinity purified
  • Storage Buffer:
  • PBS, 50% glycerol, 0.5% BSA, 0.02% sodium azide, pH 7.4
  • PBS, 50% glycerol, 0.5% BSA, 0.02% sodium azide, pH 7.4
  • Storage Temperature:
  • Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
  • Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
  • Shipping Temperature:
  • Shipped on blue ice
  • Shipped on blue ice

Specifications

Frequently Bought Together