Anti-ROR2 Rabbit Polyclonal Antibody (FITC)
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: ROR2
Clonality: Polyclonal
Clone:
Conjucation: FITC
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77713-548
- 77713-547
- Antigen Symbol:
- ROR2
- ROR2
- Antigen Synonyms:
- Brachydactyly type B|ROR2_HUMAN|Neurotrophic tyrosine kinase, receptor related 2|EC 2.7.10.1|receptor-related 2|Tyrosine-protein kinase transmembrane receptor Ror2|Neurotrophic tyrosine kinase|BDB|NTRKR2|Receptor tyrosine kinase-like orphan receptor 2|MGC163394|BDB1|Tyrosine protein kinase transmembrane receptor ROR2|ROR2
- Brachydactyly type B|ROR2_HUMAN|Neurotrophic tyrosine kinase, receptor related 2|EC 2.7.10.1|receptor-related 2|Tyrosine-protein kinase transmembrane receptor Ror2|Neurotrophic tyrosine kinase|BDB|NTRKR2|Receptor tyrosine kinase-like orphan receptor 2|MGC163394|BDB1|Tyrosine protein kinase transmembrane receptor ROR2|ROR2
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- FITC (Fluorescein Isothiocyanate)
- FITC (Fluorescein Isothiocyanate)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q01974
- Q01974
- Isotype:
- IgG
- IgG
- Immunogen:
- Recombinant Human Tyrosine-protein kinase transmembrane receptor ROR2 protein (752-926AA)
- Recombinant Human Tyrosine-protein kinase transmembrane receptor ROR2 protein (752-926AA)
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together