Anti-PDE6A Rabbit Polyclonal Antibody
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: PDE6A
Clonality: Polyclonal
Clone:
Conjucation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77713-524
- 77713-525
- Antigen Symbol:
- PDE6A
- PDE6A
- Antigen Synonyms:
- PDEA|Rod cGMP specific 3' 5' cyclic phosphodiesterase alpha subunit|Phosphodiesterase 6A cGMP specific rod alpha|5''-cyclic phosphodiesterase subunit alpha|GMP PDE alpha|PDE 6 alpha|PDE V B1|PDE V-B1|Phosphodiesterase 6A|PDE6A_HUMAN|Retinal Rod Photoreceptor cGMP Phosphodiesterase alpha|Phosphodiesterase 6 alpha|Phosphodiesterase 6A alpha subunit|CGPR A|PDE 6A|Rod cGMP-specific 3''|PDE6A|GMP-PDE alpha
- PDEA|Rod cGMP specific 3' 5' cyclic phosphodiesterase alpha subunit|Phosphodiesterase 6A cGMP specific rod alpha|5''-cyclic phosphodiesterase subunit alpha|GMP PDE alpha|PDE 6 alpha|PDE V B1|PDE V-B1|Phosphodiesterase 6A|PDE6A_HUMAN|Retinal Rod Photoreceptor cGMP Phosphodiesterase alpha|Phosphodiesterase 6 alpha|Phosphodiesterase 6A alpha subunit|CGPR A|PDE 6A|Rod cGMP-specific 3''|PDE6A|GMP-PDE alpha
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- P16499
- P16499
- Isotype:
- IgG
- IgG
- Immunogen:
- Recombinant Human Rod cGMP-specific 3\\\',5\\\'-cyclic phosphodiesterase subunit alpha protein (656-860AA)
- Recombinant Human Rod cGMP-specific 3\\\',5\\\'-cyclic phosphodiesterase subunit alpha protein (656-860AA)
- Form:
- Liquid
- Liquid
- Purification:
- Antigen affinity purified
- Antigen affinity purified
- Storage Buffer:
- PBS, 0.02% sodium azide, 50% glycerol, pH 7.3
- PBS, 0.02% sodium azide, 50% glycerol, pH 7.3
- Storage Temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together