Anti-ZBTB16 Rabbit Polyclonal Antibody
New Product
Recommended dilution: WB:1:200-1:3000, IHC:1:20-1:200
Type:
Antigen: ZBTB16
Clonality: Polyclonal
Clone:
Conjucation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
- Catalog No:
- 77713-470
- 77689-263
- Antigen Symbol:
- ZBTB16
- ZBTB16
- Antigen Synonyms:
- zinc finger and BTB domain containing protein 16|lu|PLZF|ZBTB16|Zinc finger protein 145 (Kruppel like expressed in promyelocytic leukemia)|Zfp145|ZBT16_HUMAN|AI467657|Zinc finger and BTB domain-containing protein 16|Znf145|Zinc finger protein 145|Zinc finger protein PLZF|Zinc finger and BTB domain containing 16|Promyelocytic leukemia zinc finger protein
- zinc finger and BTB domain containing protein 16|lu|PLZF|ZBTB16|Zinc finger protein 145 (Kruppel like expressed in promyelocytic leukemia)|Zfp145|ZBT16_HUMAN|AI467657|Zinc finger and BTB domain-containing protein 16|Znf145|Zinc finger protein 145|Zinc finger protein PLZF|Zinc finger and BTB domain containing 16|Promyelocytic leukemia zinc finger protein
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human, Mouse
- Human, Mouse
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q05516
- Q05516
- Isotype:
- IgG
- IgG
- Western Blot:
- Yes
- Yes
- ELISA:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- Immunogen:
- Recombinant Human Zinc finger and BTB domain-containing protein 16 protein (126-420AA)
- Recombinant Human Zinc finger and BTB domain-containing protein 16 protein (126-420AA)
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Frequently Bought Together