Anti-CENPC Rabbit Polyclonal Antibody
Supplier: AFG Bioscience
New Product
Certificates
About this item
ELISA:1:2000-1:5000, IHC:1:50-1:200
Type:
Antigen: CENPC
Clonality: Polyclonal
Clone:
Conjucation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77713-424
- 77713-425
- Antigen Synonyms:
- CENP-C|CENPC_HUMAN|Centromere autoantigen C1|Interphase centromere complex protein 7|MIF2|CENPC1|Centromere protein C 1|Centromere protein C|ICEN7|hcp-4|Centromere autoantigen C|CENP-C 1
- CENP-C|CENPC_HUMAN|Centromere autoantigen C1|Interphase centromere complex protein 7|MIF2|CENPC1|Centromere protein C 1|Centromere protein C|ICEN7|hcp-4|Centromere autoantigen C|CENP-C 1
- Gene ID:
- Q03188
- Q03188
- Immunogen:
- Fusion protein of Human CENPC
- Fusion protein of Human CENPC
- Form:
- Liquid
- Liquid
- Purification:
- Antigen affinity purified
- Antigen affinity purified
- Storage Buffer:
- PBS, 0.05% sodium azide, 40% glycerol, pH 7.4
- PBS, 0.05% sodium azide, 40% glycerol, pH 7.4
- Storage Temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together