Anti-SIGMAR1 Rabbit Polyclonal Antibody
New Product
Type:
Antigen: SIGMAR1
Clonality: Polyclonal
Clone:
Conjucation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
- Catalog No:
- 77713-389
- 77713-390
- Antigen Symbol:
- SIGMAR1
- SIGMAR1
- Antigen Synonyms:
- Aging associated gene 8 protein|SRBP|SR31747-binding protein|Sigma1-receptor|sigma1 receptor|SR31747 binding protein|Aging-associated gene 8 protein|SR BP|opioid receptor, sigma 1 isoform 1|SGMR1_HUMAN|mSigmaR1|Sigmar1|AL024364|SR-BP|Sigma non-opioid intracellular receptor 1|RP23 167I12.6|hSigmaR1|Sigma1R|opioid receptor, sigma 1|SIG-1R|Sigma 1-type opioid receptor|SIG 1R
- Aging associated gene 8 protein|SRBP|SR31747-binding protein|Sigma1-receptor|sigma1 receptor|SR31747 binding protein|Aging-associated gene 8 protein|SR BP|opioid receptor, sigma 1 isoform 1|SGMR1_HUMAN|mSigmaR1|Sigmar1|AL024364|SR-BP|Sigma non-opioid intracellular receptor 1|RP23 167I12.6|hSigmaR1|Sigma1R|opioid receptor, sigma 1|SIG-1R|Sigma 1-type opioid receptor|SIG 1R
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human, Rat, Mouse
- Human, Rat, Mouse
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q99720
- Q99720
- Isotype:
- IgG
- IgG
- Immunogen:
- Fusion protein of Human SIGMAR1
- Fusion protein of Human SIGMAR1
- Form:
- Liquid
- Liquid
- Purification:
- Antigen affinity purified
- Antigen affinity purified
- Storage Buffer:
- PBS, 0.05% sodium azide, 40% glycerol, pH 7.4.
- PBS, 0.05% sodium azide, 40% glycerol, pH 7.4.
- Storage Temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Frequently Bought Together