Anti-ANXA3 Rabbit Polyclonal Antibody
Supplier: AFG Bioscience
New Product
Certificates
About this item
ELISA:1:2000-1:5000, WB:1:500-1:2000, IHC:1:50-1:200
Type:
Antigen: ANXA3
Clonality: Polyclonal
Clone:
Conjucation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Specifications
- Catalog No:
- 77712-790
- 77712-789
- Antigen Symbol:
- ANXA3
- ANXA3
- Antigen Synonyms:
- OTTHUMP00000219237|Placental anticoagulant protein III|ANXA 3|Annexin A3|ANX3|Lipocortin III|Annexin III|35-alpha calcimedin|OTTHUMP00000219238|PAP III|Calcimedin 35 alpha|PAP-III|2-cyclic phosphate 2-phosphohydrolase|Anxa3|ANXA3_HUMAN|Inositol 1 2 cyclic phosphate 2 phosphohydrolase |Annexin-3|OTTHUMP00000160701|Inositol 1
- OTTHUMP00000219237|Placental anticoagulant protein III|ANXA 3|Annexin A3|ANX3|Lipocortin III|Annexin III|35-alpha calcimedin|OTTHUMP00000219238|PAP III|Calcimedin 35 alpha|PAP-III|2-cyclic phosphate 2-phosphohydrolase|Anxa3|ANXA3_HUMAN|Inositol 1 2 cyclic phosphate 2 phosphohydrolase |Annexin-3|OTTHUMP00000160701|Inositol 1
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human, Rat, Mouse
- Human, Rat, Mouse
- Host:
- Rabbit
- Rabbit
- Gene ID:
- P12429
- P12429
- Isotype:
- IgG
- IgG
- Western Blot:
- Yes
- Yes
- ELISA:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- Immunogen:
- Fusion protein of Human ANXA3
- Fusion protein of Human ANXA3
- Form:
- Liquid
- Liquid
- Purification:
- Antigen affinity purified
- Antigen affinity purified
- Storage Buffer:
- PBS, 0.05% sodium azide, 40% glycerol, pH 7.4.
- PBS, 0.05% sodium azide, 40% glycerol, pH 7.4.
- Storage Temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together