Anti-NDUFB5 Rabbit Polyclonal Antibody
Catalog # 77688-324
Supplier: AFG Bioscience
New Product
Specifications
- Antigen Symbol:NDUFB5
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human
- Western Blot:Yes
- Size:100 µg
- Form:Liquid
- Gene ID:O43674
- Antigen Synonyms:CI SGDH|NADH ubiquinone oxidoreductase SGDH subunit|MGC12314|NADH dehydrogenase [ubiquinone] 1 beta subcomplex subunit 5 mitochondrial|NDUB5_HUMAN|Complex I-SGDH|NADH dehydrogenase (ubiquinone) 1 beta subcomplex 5|NADH-ubiquinone oxidoreductase SGDH subunit|SGDH|NADH dehydrogenase (ubiquinone) 1 beta subcomplex 5 16kDa|mitochondrial|NADH dehydrogenase [ubiquinone] 1 beta subcomplex subunit 5|FLJ30597|Complex I SGDH|CI-SGDH|MGC111204|DKFZp686N02262|Ndufb5
- Storage Buffer:0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on blue ice
- Immunogen:Recombinant Human NADH dehydrogenase [ubiquinone] 1 beta subcomplex subunit 5, mitochondrial protein (94-189AA)
- Purification:>95%, Protein G purified
- Cat. No.:77688-324
- Packaging:Vial
Specifications
About this item
Recommended dilution: WB:1:500-1:5000
Type:
Antigen: NDUFB5
Clonality: Polyclonal
Clone:
Conjucation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human