Anti-MAGEA3 Rabbit Polyclonal Antibody
77688-210
New Product
- Antigen Symbol:MAGEA3
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human
- Western Blot:Yes
- Size:100 µg
- Form:Liquid
- Gene ID:P43357
- Antigen Synonyms:Antigen MZ2-D|Melanoma associated antigen 3|CT1.3|MAGEA6|HIP8|MAGE family member A3|MAGE3|Melanoma antigen family A 3|Cancer/testis antigen family 1 member 3|Cancer/testis antigen 1.3|MAGA3_HUMAN|MAGE-3 antigen|Melanoma-associated antigen 3|HYPD|MGC1461|Antigen MZ2 D |MAGE 3 antigen|MAGEA3
- Storage Buffer:0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on blue ice
- Immunogen:Recombinant Human Melanoma-associated antigen 3 protein (2-104AA)
- Purification:>95%, Protein G purified
- Cat. No.:77688-210
- Packaging:Vial
Recommended dilution: WB:1:500-1:5000, IHC:1:200-1:500
Type:
Antigen: MAGEA3
Clonality: Polyclonal
Clone:
Conjucation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human