Anti-JUND Rabbit Polyclonal Antibody
77703-380
New Product
- Antigen Symbol:JUND
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoPrecipitation:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western Blot:Yes
- Size:100 µg
- Form:Liquid
- Gene ID:P17535
- Antigen Synonyms:JunD FL isoform|Jun D|JUND_HUMAN|AP1|Jund|jun D proto oncogene|Transcription factor jun D|Transcription factor jun-D|AP 1|Activator protein 1
- Storage Buffer:PBS, 50% glycerol, 0.5% BSA, 0.02% sodium azide, pH 7.4.
- Storage Temperature:Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on blue ice
- Immunogen:Synthesized peptide derived from Human Jun D around the non-phosphorylation site of S255.
- Purification:Antigen affinity purified
- Cat. No.:77703-380
- Packaging:Vial
WB:1:500-1:2000, IHC:1:100-1:300, IP:2-5ug, ELISA:1:20000
Type:
Antigen: JUND
Clonality: Polyclonal
Clone:
Conjucation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat