Order Entry
United States
ContactUsLinkComponent
Ag, Al, As, Ba, Be, Bi, Ca, Cd, Co, Cr, Cs, Cu, Fe, Ga, In, K, Li, Mg, Mn, Na, Ni, Pb, Rb, Se, Sr, Tl, U, V, Zn @ 10 ug/mL in 5% HNO3, VHG, LGC Standards
Ag, Al, As, Ba, Be, Bi, Ca, Cd, Co, Cr, Cs, Cu, Fe, Ga, In, K, Li, Mg, Mn, Na, Ni, Pb, Rb, Se, Sr, Tl, U, V, Zn @ 10 ug/mL in 5% HNO3, VHG, LGC Standards
Catalog # 77697-853
Supplier:  LGC Standards
New Product
CAS Number:  
undefined
Ag, Al, As, Ba, Be, Bi, Ca, Cd, Co, Cr, Cs, Cu, Fe, Ga, In, K, Li, Mg, Mn, Na, Ni, Pb, Rb, Se, Sr, Tl, U, V, Zn @ 10 ug/mL in 5% HNO3, VHG, LGC Standards
Catalog # 77697-853
Supplier:  LGC Standards
Supplier Number:  VHG-SM29-1-100
New Product
CAS Number:  

Specifications

  • Concentration:
    10 µg/mL
  • Matrix:
    5% HNO₃
  • Description:
    Ag, Al, As, Ba, Be, Bi, Ca, Cd, Co, Cr, Cs, Cu, Fe, Ga, In, K, Li, Mg, Mn, Na, Ni, Pb, Rb, Se, Sr, Tl, U, V, Zn @ 10 ug/mL in 5% HNO3
  • Size:
    100 ml
  • Cat. No.:
    77697-853

Specifications

About this item