K @ 1000; Ag, Al, As, B, Ba, Be, Ca, Cd, Co, Cu, Fe, Mg, Mn, Mo, Na, Ni, Pb, Sb, Se, Ti, Tl, V, Zn @ 100; Si @ 50 ug/mL in 5% HNO3/tr. HF, VHG, LGC Standards
Catalog # 77697-845
Supplier: LGC Standards
New Product
CAS Number:
Specifications
- Concentration:Varies
- Matrix:5% HNO₃/tr HF
- Description:K @ 1000; Ag, Al, As, B, Ba, Be, Ca, Cd, Co, Cu, Fe, Mg, Mn, Mo, Na, Ni, Pb, Sb, Se, Ti, Tl, V, Zn @ 100; Si @ 50 ug/mL in 5% HNO3/tr. HF
- Size:100 ml
- Cat. No.:77697-845