QC Standard 7A: Ag @ 50; Al, B, Ba, Na @ 100; Si @ 500; K @ 1000 µg/mL in 5% HNO3, tr. F-, VHG, LGC Standards
Catalog # 77697-844
Supplier: LGC Standards
New Product
CAS Number:
Specifications
- Concentration:Varies
- Matrix:5% HNO₃/tr HF
- Description:QC Standard 7A: Ag @ 50; Al, B, Ba, Na @ 100; Si @ 500; K @ 1000 µg/mL in 5% HNO3, tr. F-
- Size:100 ml
- Cat. No.:77697-844